Learn More
Aviva Systems Biology Corp MARCH3 Antibody - C-terminal region (ARP43464_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43464_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against March3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 6330411I15Rik, A530081L18Rik, AI843639, MARCH-III, MGC130167, MGC130168ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-074