Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Marchantia, Living Material Coupon
SDPSCI_ED

Catalog No. S37287 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Sufficient For:
Class of 30 students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
S37287 Class of 30 students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37287 Supplier Carolina™ Supplier No. 156540LC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as pre-paid coupon to order living material at a future date. Carolina Marchantia, Living Material Coupon is not live material.

  • Liverwort most often described in textbooks
  • Each portion equals 11.4 x 11.4cm (4.5 x 4.5 in.)

Specifications

Quantity 1 Ea.
Sufficient For Class of 30 students
Product Type Marchantia, Living Material Coupon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.