Learn More
Aviva Systems Biology Corp MARCKS Antibody - C-terminal region (ARP60325_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60325_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MARCKS. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MARCKS. Alternate names: 80K-L, FLJ14368, FLJ90045, MACS, PKCSL, PRKCSLApplicationWB, IHC. Species reactivity: Cow, Human, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-007