Learn More
Enzo Life Sciences MARCKS protein (bovine brain) (151-175) (1mg). CAS: 125267-21-2
SDP
Supplier:  Enzo Life Sciences ALX162004M001
Encompass_Preferred
Excellent substrate for protein kinase C (PKC) (Km = 20nM for purified rat brain PKC). The peptide consists of residues 151-175 of the myristoylated alanine-rich C-kinase substrate (MARCKS) protein phosphorylation site. Protein kinase C isotypes (including PKCε) phosphorylate serines 8, 12 and 19 of this peptide. It is a very weak substrate for cAMP- and cGMP-dependent protein kinase and CaM kinases I, II and III. Formula: C147H243N41O31. MW: 3081.8. Purity: ≥98%. Appearance: White to off-white powder. . Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7829