Learn More
Aviva Systems Biology Corp MARCO Antibody - N-terminal region (ARP41580_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41580_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MARCO. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MARCO. Alternate names: SCARA2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57914