Learn More
Biomatik Corporation Marco Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10867
Encompass_Preferred
Marco Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MARCO. The Marco Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Macrophage Receptor Marco Protein (421-520Aa). Applications: ELISA, WB, IHC. Target Synonyms: MARCO. Accession Number: Q9UEW3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5468