Learn More
Medchemexpress LLC HY-50098 5mg Medchemexpress, Mardepodect CAS:898562-94-2 Purity:>98%
SDP
Catalog No. 501970068
Encompass_Preferred
Medchemexpress, HY-50098 5mg Mardepodect CAS:898562-94-2 Mardepodect (PF-2545920) is a potent, orally active and selective PDE10A inhibitor with an IC50 of 0.37 nM, with >1000-fold selectivity over other PDEs. Mardepodect can cross the blood-brain barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0068 Supplier Medchemexpress LLC Supplier No. HY500985MG
Medchemexpress, HY-50098 5mg Mardepodect CAS:898562-94-2 Mardepodect (PF-2545920) is a potent, orally active and selective PDE10A inhibitor with an IC50 of 0.37 nM, with >1000-fold selectivity over other PDEs. Mardepodect can cross the blood-brain barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.