Learn More
Selleck Chemical LLC Mardepodect (PF-2545920) S2687-5mg
SDP
Catalog No. 503112629
Encompass_Preferred
Mardepodect (PF-2545920) is a potent and selective PDE10A inhibitor with IC50 of 0 37 nM with 1000-fold selectivity over the PDE
Catalog No. 50-311-2629 Supplier Selleck Chemical LLC Supplier No. S26875MG
Mardepodect (PF-2545920) is a potent and selective PDE10A inhibitor with IC50 of 0 37 nM with 1000-fold selectivity over the PDE