Learn More
Medchemexpress LLC Mardepodect hydrochloride | 2070014-78-5 | 99.9% | 10 MG
SDP
Catalog No. 5000298760
Encompass_Preferred
Mardepodect hydrochloride is the hydrochloride salt of mardepodect (PF-2545920), a potent and selective phosphodiesterase 10A (PDE10A) inhibitor used in neuroscience research. It is supplied as a solid hydrochloride salt with high reported purity and is used for in vitro and in vivo studies of PDE10A-related signaling and CNS pharmacology.
  • Potent, selective PDE10A inhibitor (reported IC50 = 0.37 nM).
  • High reported purity, suitable for research applications.
  • Hydrochloride salt form for improved solubility compared with the free base.
  • Known to cross the blood-brain barrier, relevant for CNS studies.
  • Chemical formula C25H21ClN4O and molecular weight 428.91.
  • Provided as a small solid quantity appropriate for early-stage experiments.
Catalog No. 50-002-98760 Supplier Medchemexpress LLC Supplier No. HY50098A10MG
Mardepodect hydrochloride is the hydrochloride salt of mardepodect (PF-2545920), a potent and selective phosphodiesterase 10A (PDE10A) inhibitor used in neuroscience research. It is supplied as a solid hydrochloride salt with high reported purity and is used for in vitro and in vivo studies of PDE10A-related signaling and CNS pharmacology.
  • Potent, selective PDE10A inhibitor (reported IC50 = 0.37 nM).
  • High reported purity, suitable for research applications.
  • Hydrochloride salt form for improved solubility compared with the free base.
  • Known to cross the blood-brain barrier, relevant for CNS studies.
  • Chemical formula C25H21ClN4O and molecular weight 428.91.
  • Provided as a small solid quantity appropriate for early-stage experiments.