Learn More
Medchemexpress LLC Mardepodect hydrochloride | 2070014-78-5 | 99.9% | C25H21ClN4O | 25 MG
SDP
Catalog No. 5000359123
Encompass_Preferred
Mardepodect hydrochloride is a potent, orally active, and selective PDE10A inhibitor with an IC50 of 0.37 nM, demonstrating over 1000-fold selectivity compared to other PDEs. It is capable of crossing the blood-brain barrier.
  • Potent, orally active, and selective PDE10A inhibitor
  • IC50 of 0.37 nM
  • Exhibits >1000-fold selectivity over other PDEs
  • Can cross the blood-brain barrier
  • Active in the conditioned avoidance response (CAR) assay with an ED50 of 1 mg/kg
  • Induces a dose-dependent increase in striatal cGMP in mice
Catalog No. 50-003-59123 Supplier Medchemexpress LLC Supplier No. 25MGHY50098A
Mardepodect hydrochloride is a potent, orally active, and selective PDE10A inhibitor with an IC50 of 0.37 nM, demonstrating over 1000-fold selectivity compared to other PDEs. It is capable of crossing the blood-brain barrier.
  • Potent, orally active, and selective PDE10A inhibitor
  • IC50 of 0.37 nM
  • Exhibits >1000-fold selectivity over other PDEs
  • Can cross the blood-brain barrier
  • Active in the conditioned avoidance response (CAR) assay with an ED50 of 1 mg/kg
  • Induces a dose-dependent increase in striatal cGMP in mice