Learn More
TARGETMOL CHEMICALS INC MAREIN 5MG
SDP
Catalog No. 502576170
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Marein shows neuroprotective effect on PC12 cell damage induced by methylglyoxal which is due to a reduction of damage to mitochondria function and activation of the AMPK signal pathway it may be a potent compound for preventing/counteracting diabetic encephalopathy. purity: 99%
Catalog No. 50-257-6170 Supplier TARGETMOL CHEMICALS INC Supplier No. TN19115MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Marein shows neuroprotective effect on PC12 cell damage induced by methylglyoxal which is due to a reduction of damage to mitochondria function and activation of the AMPK signal pathway it may be a potent compound for preventing/counteracting diabetic encephalopathy. purity: 99%