Learn More
Cayman Chemical MaresIn 1-d5 10ug
SDP
Catalog No. 502986080
Encompass_Preferred
An internal standard for the quantification of maresin 1 by GC- or LC-MS
Catalog No. 50-298-6080 Supplier Cayman Chemical Supplier No. 2182310 UG
An internal standard for the quantification of maresin 1 by GC- or LC-MS