Learn More
Cayman Chemical MaresIn 2-d5 10ug
SDP
Catalog No. 502982325
Encompass_Preferred
An internal standard for the quantification of MaR2 by GC- or LC-MS
Catalog No. 50-298-2325 Supplier Cayman Chemical Supplier No. 2075710 UG
An internal standard for the quantification of MaR2 by GC- or LC-MS