Learn More
Medchemexpress LLC Margatoxin | 145808-47-5 | 99.17% | 4178.95 | 1 MG
SDP
Catalog No. 5000422319
Encompass_Preferred
Margatoxin, an alpha-KTx scorpion toxin, is a high-affinity inhibitor of Kv1.3 (Kd=11.7 pM). This 39 amino-acid-long peptide is isolated from the venom of the scorpion *Centruroides margaritatus* and is widely used in ion channel research.
  • High affinity inhibitor of Kv1.3, Kv1.2, and Kv1.1
  • Isolated from *Centruroides margaritatus* scorpion venom
  • Used in ion channel research
  • Decreases thioglycollate-induced leukocyte transmigration in mice
Catalog No. 50-004-22319 Supplier Medchemexpress LLC Supplier No. HYP12801MG
Margatoxin, an alpha-KTx scorpion toxin, is a high-affinity inhibitor of Kv1.3 (Kd=11.7 pM). This 39 amino-acid-long peptide is isolated from the venom of the scorpion *Centruroides margaritatus* and is widely used in ion channel research.
  • High affinity inhibitor of Kv1.3, Kv1.2, and Kv1.1
  • Isolated from *Centruroides margaritatus* scorpion venom
  • Used in ion channel research
  • Decreases thioglycollate-induced leukocyte transmigration in mice