Learn More
Medchemexpress LLC Margatoxin | 145808-47-5 | 99.2% | 4178.95 | 100 UG
SDP
Catalog No. 5000422996
Encompass_Preferred
Margatoxin is an alpha-KTx scorpion toxin, a 39 amino acid peptide isolated from the venom of the Centruroides margaritatus scorpion. It functions as a high-affinity inhibitor of Kv1.3, Kv1.2, and Kv1.1 potassium channels, making it a valuable tool in ion channel research.
  • High-affinity inhibitor of Kv1.3, Kv1.2, and Kv1.1 potassium channels
  • 39 amino acid peptide
  • Derived from *Centruroides margaritatus* scorpion venom
  • Used in ion channel research
  • Decreases thioglycollate-induced leukocyte transmigration in vivo
Catalog No. 50-004-22996 Supplier Medchemexpress LLC Supplier No. HYP1280100UG
Margatoxin is an alpha-KTx scorpion toxin, a 39 amino acid peptide isolated from the venom of the Centruroides margaritatus scorpion. It functions as a high-affinity inhibitor of Kv1.3, Kv1.2, and Kv1.1 potassium channels, making it a valuable tool in ion channel research.
  • High-affinity inhibitor of Kv1.3, Kv1.2, and Kv1.1 potassium channels
  • 39 amino acid peptide
  • Derived from *Centruroides margaritatus* scorpion venom
  • Used in ion channel research
  • Decreases thioglycollate-induced leukocyte transmigration in vivo