Learn More
Medchemexpress LLC Margatoxin | 145808-47-5 | 99.2% | 4178.95 | 500 UG
SDP
Catalog No. 5000423654
Encompass_Preferred
Margatoxin is an alpha-KTx scorpion toxin and a high-affinity inhibitor of Kv1.3 (Kd=11.7 pM), Kv1.2 (Kd=6.4 pM), and Kv1.1 (Kd=4.2 nM). This 39 amino-acid-long peptide is isolated from the venom of the scorpion Centruroides margaritatus and is widely used in ion channel research. It is supplied as a white to off-white solid and has been shown to significantly decrease thioglycollate-induced leukocyte transmigration in mice.
  • High-affinity inhibitor of Kv1.3, Kv1.2, and Kv1.1
  • 39 amino-acid-long peptide
  • Isolated from Centruroides margaritatus venom
  • Used extensively in ion channel research
  • Decreases leukocyte transmigration in vivo
Catalog No. 50-004-23654 Supplier Medchemexpress LLC Supplier No. HYP1280500UG
Margatoxin is an alpha-KTx scorpion toxin and a high-affinity inhibitor of Kv1.3 (Kd=11.7 pM), Kv1.2 (Kd=6.4 pM), and Kv1.1 (Kd=4.2 nM). This 39 amino-acid-long peptide is isolated from the venom of the scorpion Centruroides margaritatus and is widely used in ion channel research. It is supplied as a white to off-white solid and has been shown to significantly decrease thioglycollate-induced leukocyte transmigration in mice.
  • High-affinity inhibitor of Kv1.3, Kv1.2, and Kv1.1
  • 39 amino-acid-long peptide
  • Isolated from Centruroides margaritatus venom
  • Used extensively in ion channel research
  • Decreases leukocyte transmigration in vivo