Learn More
Medchemexpress LLC Maribavir | 176161-24-3 | 99.96% | 376.24 | 1 ML
SDP
Catalog No. 5000394998
Encompass_Preferred
Maribavir is a potent inhibitor of histone phosphorylation catalyzed by wild-type pUL97 in vitro, with an IC50 of 3 nM. It also demonstrates potent antiviral activity against HCMV and Epstein-Barr virus (EBV).
  • Potent inhibitor of histone phosphorylation.
  • Demonstrates potent antiviral activity against HCMV and Epstein-Barr virus (EBV).
  • For research use only.
  • Multiple forms available.
Catalog No. 50-003-94998 Supplier Medchemexpress LLC Supplier No. HY1630510MM/1ML
Maribavir is a potent inhibitor of histone phosphorylation catalyzed by wild-type pUL97 in vitro, with an IC50 of 3 nM. It also demonstrates potent antiviral activity against HCMV and Epstein-Barr virus (EBV).
  • Potent inhibitor of histone phosphorylation.
  • Demonstrates potent antiviral activity against HCMV and Epstein-Barr virus (EBV).
  • For research use only.
  • Multiple forms available.