Learn More
Medchemexpress LLC Maribavir-d6 | 176161-24-3 | 99.8% | 382.27 | 1 MG
SDP
Catalog No. 5000394940
Encompass_Preferred
Maribavir-d6 (1263W94-d6) is a deuterium labeled Maribavir (HY-16305). Maribavir is a potent inhibitor of histone phosphorylation catalyzed by wild-type pUL97 in vitro, with an IC50 of 3 nM.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-94940 Supplier Medchemexpress LLC Supplier No. HY16305S1MG
Maribavir-d6 (1263W94-d6) is a deuterium labeled Maribavir (HY-16305). Maribavir is a potent inhibitor of histone phosphorylation catalyzed by wild-type pUL97 in vitro, with an IC50 of 3 nM.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.