Learn More
Medchemexpress LLC Maribavir-d6 | 176161-24-3 | 99.8% | 382.27 | 5 MG
SDP
Catalog No. 5000394332
Encompass_Preferred
Maribavir-d6 is a deuterium-labeled Maribavir, a potent inhibitor of histone phosphorylation catalyzed by wild-type pUL97 in vitro, with an IC50 of 3 nM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-94332 Supplier Medchemexpress LLC Supplier No. HY16305S5MG
Maribavir-d6 is a deuterium-labeled Maribavir, a potent inhibitor of histone phosphorylation catalyzed by wild-type pUL97 in vitro, with an IC50 of 3 nM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.