Learn More
Medchemexpress LLC Maribavir-d6 TFA | 176161-24-3 | 496.30 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY16305S11MG
Encompass_Preferred
Maribavir-d6 TFA is a deuterium-labeled Maribavir, an orally active antiviral agent. It targets cytomegalovirus (CMV) by inhibiting protein kinase UL97, thereby preventing CMV DNA replication. This compound is suitable for various analytical techniques.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-94634