Learn More
Electron Microscopy Sciences Marienfeld Superior™ Precision Cover Glass, Thickness No. 1.5H (tol ± 5 µm) for high-performance microscope objectives. This high-precision cover glass features exceptionally accurate thickness of 170 µm ± 5 µm.
SDP
Catalog No. NC3832788
Encompass_Preferred
Marienfeld Superior™ Precision Cover Glass, Thickness No. 1.5H (tol ± 5 µm) for high-performance microscope objectives. Compliance with the nominal thickness has a critical impact on imaging quality. Thickness variations reduce image contrast and produce image defects, particularly spherical aberrations. This high-precision cover glass with thickness No. 1.5H features exceptionally accurate thickness of 170 µm ± 5 µm.
Catalog No. NC3832788 Supplier Electron Microscopy Sciences Supplier No. 71861054
Marienfeld Superior™ Precision Cover Glass, Thickness No. 1.5H (tol ± 5 µm) for high-performance microscope objectives. Compliance with the nominal thickness has a critical impact on imaging quality. Thickness variations reduce image contrast and produce image defects, particularly spherical aberrations. This high-precision cover glass with thickness No. 1.5H features exceptionally accurate thickness of 170 µm ± 5 µm.