Learn More
Medchemexpress LLC HY-12169 50mg , Marimastat BB2516;TA2516 CAS:154039-60-8 Purity:98%
SDP
Catalog No. 501493721
Encompass_Preferred
Medchemexpress, HY-12169 50mg Marimastat BB2516;TA2516 CAS:154039-60-8 Formula:C15H29N3O5 IC50: 3 nM (MMP-9), 5 nM (MMP-1), 6 nM (MMP-2), 9 nM (MMP-14), 13 nM (MMP-7) Purity:98% Marimastat is a broad spectrum inhibitor of MMPs with IC 50 values of 3, 5, 6, 9 and 13 nM for MMP-9, MMP-1, MMP-2, MMP-14 and MMP-7, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3721 Supplier Medchemexpress LLC Supplier No. HY1216950MG
Medchemexpress, HY-12169 50mg Marimastat BB2516;TA2516 CAS:154039-60-8 Formula:C15H29N3O5 IC50: 3 nM (MMP-9), 5 nM (MMP-1), 6 nM (MMP-2), 9 nM (MMP-14), 13 nM (MMP-7) Purity:98% Marimastat is a broad spectrum inhibitor of MMPs with IC 50 values of 3, 5, 6, 9 and 13 nM for MMP-9, MMP-1, MMP-2, MMP-14 and MMP-7, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.