Learn More
Apexbio Technology LLC Marimastat, 1mg. Cas: 154039-60-8 MFCD: MFCD00866242. MMPs inhibitor,board spectrum
SDP
Supplier:  Apexbio Technology LLC A40491
Encompass_Preferred
Marimastat is a broad range inhibitor of Matrix metalloproteinases with IC50 values of 5, 6, 13, 3 and 9 nM for MMP-1, MMP-2, MMP-7 , MMP-9 and MMP-14 [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4866
 Restricted Item