Learn More
Apexbio Technology LLC Marimastat 154039-60-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064385
Encompass_Preferred
Marimastat (CAS 154039-60-8) also known as BB-2516 is an orally bioavailable inhibitor of matrix metalloproteases (MMPs) Structurally it contains a collagen-mimicking hydroxamate moiety that facilitates covalent binding to the zinc atom at MMP active sites Marimastat exhibits broad-spectrum inhibitory activity against MMP-1 MMP-2 MMP-7 MMP-9 and MMP-14 with IC50 values of 5 nM 6 nM 13 nM 3 nM and 9 nM respectively In animal models of metastatic cancers Marimastat reduces tumor dissemination and progression Clinically Marimastat is investigated for potential therapeutic effects against metastatic and non-metastatic solid tumors such as pancreatic and gastric cancers
Catalog No. 50-000-64385 Supplier Apexbio Technology LLC Supplier No. A40495.1
Marimastat (CAS 154039-60-8) also known as BB-2516 is an orally bioavailable inhibitor of matrix metalloproteases (MMPs) Structurally it contains a collagen-mimicking hydroxamate moiety that facilitates covalent binding to the zinc atom at MMP active sites Marimastat exhibits broad-spectrum inhibitory activity against MMP-1 MMP-2 MMP-7 MMP-9 and MMP-14 with IC50 values of 5 nM 6 nM 13 nM 3 nM and 9 nM respectively In animal models of metastatic cancers Marimastat reduces tumor dissemination and progression Clinically Marimastat is investigated for potential therapeutic effects against metastatic and non-metastatic solid tumors such as pancreatic and gastric cancers