Learn More
Selleck Chemical LLC Marimastat
SDP
Catalog No. 5000002236
Encompass_Preferred
Marimastat (BB-2516 TA2516) is a broad spectrum matrix metalloprotease (MMP) inhibitor for MMP-9 MMP-1 MMP-2 MMP-14 and MMP-7 with IC50 of 3 nM 5 nM 6 nM 9 nM and 13 nM respectively Phase 3
Catalog No. 50-000-02236 Supplier Selleck Chemical LLC Supplier No. S715625MG
Marimastat (BB-2516 TA2516) is a broad spectrum matrix metalloprotease (MMP) inhibitor for MMP-9 MMP-1 MMP-2 MMP-14 and MMP-7 with IC50 of 3 nM 5 nM 6 nM 9 nM and 13 nM respectively Phase 3