Learn More
Selleck Chemical LLC Marimastat (BB-2516) 5mg 154039-60-8
SDP
Catalog No. NC1107259
Encompass_Preferred
Marimastat (BB-2516) is a broad spectrum matrix metalloprotease (MMP) inhibitor for MMP-9, MMP-1, MMP-2, MMP-14 and MMP-7 with IC50 of 3 nM, 5 nM, 6 nM, 9 nM and 13 nM, respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1107259 Supplier Selleck Chemical LLC Supplier No. S71565MG
Marimastat (BB-2516) is a broad spectrum matrix metalloprotease (MMP) inhibitor for MMP-9, MMP-1, MMP-2, MMP-14 and MMP-7 with IC50 of 3 nM, 5 nM, 6 nM, 9 nM and 13 nM, respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.