Learn More
Selleck Chemical LLC Marinopyrrole A (Maritoclax) 5mg 1227962-62-0
SDP
Catalog No. 501948237
Encompass_Preferred
Marinopyrrole A (Maritoclax) is a selective Mcl-1 antagonist. It binds to Mcl-1, but not Bcl-XL, and targets Mcl-1 for proteasomal degradation. Maritoclax disrupts the interaction between Bim and Mcl-1 with an IC50 of 10.1 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8237 Supplier Selleck Chemical LLC Supplier No. S71265MG
Marinopyrrole A (Maritoclax) is a selective Mcl-1 antagonist. It binds to Mcl-1, but not Bcl-XL, and targets Mcl-1 for proteasomal degradation. Maritoclax disrupts the interaction between Bim and Mcl-1 with an IC50 of 10.1 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.