Learn More
MSC Mark-10 Digital Force Gage, 200 lb Capacity, 0.2 lb Resolution, NIST Traceable
Catalog No. 5000527765
Encompass
The Mark-10 M3-200 Digital Force Gage is designed for precise measurements of compression and tension, featuring a high sampling rate of 2000 Hz for accurate data capture. It offers a 200 lb capacity with a 0.2 lb resolution, making it suitable for various laboratory and industrial applications requiring reliable force measurement.
  • Tip types include hook, V-groove, flat, chisel, and conical.
  • Constructed with an aluminum housing for durability.
  • Can be powered by AC or battery, with NiMH 8.4V batteries included.
  • Features peak mode for capturing maximum force readings.
  • Includes a NIST-traceable certificate of calibration.
  • Comes with a quick-start guide, resource CD, and USB cable.
  • Supplied with a cushioned carrying case for protection and portability.
  • Universal voltage AC adapter and extension rod are included.
Catalog No. 50-005-27765 Supplier MSC Supplier No. 55173454
The Mark-10 M3-200 Digital Force Gage is designed for precise measurements of compression and tension, featuring a high sampling rate of 2000 Hz for accurate data capture. It offers a 200 lb capacity with a 0.2 lb resolution, making it suitable for various laboratory and industrial applications requiring reliable force measurement.
  • Tip types include hook, V-groove, flat, chisel, and conical.
  • Constructed with an aluminum housing for durability.
  • Can be powered by AC or battery, with NiMH 8.4V batteries included.
  • Features peak mode for capturing maximum force readings.
  • Includes a NIST-traceable certificate of calibration.
  • Comes with a quick-start guide, resource CD, and USB cable.
  • Supplied with a cushioned carrying case for protection and portability.
  • Universal voltage AC adapter and extension rod are included.