Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MARK-IN-4 | 1990492-86-8 | 99.6% | 421.52 | 1 ML
SDP

Catalog No. 5000438175
Encompass_Preferred

MARK-IN-4 (Compound 18) is a potent MARK inhibitor with an IC50 of 1 nM, making it suitable for Alzheimer's disease research.

  • Potent MARK inhibitor (IC50 of 1 nM)
  • Applicable to Alzheimer's disease research
  • High purity: 99.63%
  • CAS number: 1990492-86-8
  • Molecular weight: 421.52
  • Appearance: Solid, light yellow to yellow
  • Recommended storage: -20°C, protect from light, stored under nitrogen

Catalog No. 50-004-38175 Supplier Medchemexpress LLC Supplier No. HY11226610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MARK-IN-4 (Compound 18) is a potent MARK inhibitor with an IC50 of 1 nM, making it suitable for Alzheimer's disease research.

  • Potent MARK inhibitor (IC50 of 1 nM)
  • Applicable to Alzheimer's disease research
  • High purity: 99.63%
  • CAS number: 1990492-86-8
  • Molecular weight: 421.52
  • Appearance: Solid, light yellow to yellow
  • Recommended storage: -20°C, protect from light, stored under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.