Learn More
Epredia™ Mark-It™ Tissue Marking Dyes
Catalog No. 22050456 Shop All Epredia Products
Change view
Click to view available options
Black
Catalog No.
22-050-456 Black
1 options
This item is not returnable. View return policy
Catalog No. 22-050-456 Supplier Epredia™ Supplier No. 5000BK
Add to Cart
Add to Cart
This item is not returnable. View return policy
For a variety of tissue identification purposes use Epredia™ Mark-It™ Tissue Marking Dyes.
Features:
  • Permanent dyes that maintain their original brilliance during routine tissue processing cycles
  • Can be used on fresh or formalin-fixed tissues
Uses include:
  • Marking surgical margins
  • Identifying serial segments of large tissues to ensure proper orientation and embedding
  • Maintaining proper tissue orientation for Mohs surgery specimens
  • Identification of multiple pieces of tissues processed in the same cassette
Ordering Information:
  • Available in black, blue, green, orange, red and yellow in 2 oz. bottles
WARNING: Cancer - www.P65Warnings.ca.gov