Learn More
Aviva Systems Biology Corp MARK3 Antibody - middle region (ARP56343_P050) | 100uL
SDP
Catalog No. 5001461821
Encompass_Preferred
This is a rabbit polyclonal antibody against MARK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MARK3. Alternate names: CTAK1, KP78, PAR1A, Par-1aApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61821 Supplier Aviva Systems Biology Corp Supplier No. ARP56343_P050
This is a rabbit polyclonal antibody against MARK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MARK3. Alternate names: CTAK1, KP78, PAR1A, Par-1aApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish