Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MARK3, GST Tag Recombinant Protein

Catalog No. PV4820
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4820 1 mg
PR8905A 100 μg
PV4819 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV4820 Supplier Invitrogen™ Supplier No. PV4820
Add to Cart
Add to Cart
Request a Quote

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

MARK3 is a member of a group of kinases known to regulate cell polarity via phosphorylation of microtubule-associated proteins (MAPs). The protein encoded by MARK3 is activated by phosphorylation and in turn is involved in the phosphorylation of tau proteins MAP2 and MAP4. Several transcript variants encoding different isoforms have been found for MARK3.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27448
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.02% Triton X-100, 50% glycerol, 2mM DTT, 150mM NaCl, 0.5mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 4140
Molecular Weight (g/mol) 108.9 kDa
Name Human MARK3, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1600015G02Rik; A430080F22Rik; cdc25C-associated protein kinase 1; cTAK1; C-TAK1; ELKL motif kinase 2; Emk2; EMK-2; EMK2/MARK3 serine-threonine protein kinase; ETK1; ETK-1; KP78; MAP/microtubule affinity regulating kinase 3; MAP/microtubule affinity-regulating kinase 3; MARK3; microtubule affinity regulating kinase 3; Mpk10; MPK-10; PAR1A; Par-1a; Protein kinase STK10; ser/Thr protein kinase PAR-1; Serine/threonine-protein kinase p78
Common Name MARK3
Gene Symbol MARK3
Kinase Family CAMKL Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.02% Trition X-100, 0.5 mM EDTA, 2 mM DTT, 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Kinase Group Ser⁄Thr Kinases: CAMK Group
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length
Form Liquid
Research Category Drug Development
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.