Learn More
Carnabio Usa Inc MARK4/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 021235
Encompass
MARK4, Full-length, Wild type, Amino Acid 1-688, NP_113605.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0511