Learn More
Microsolv Technology Corporation Marker, Laboratory, Semi - Permanent. 1.5mm Line. 13ml, Blue Ink. 4/PK.
Catalog No. 502270499
Encompass_Preferred
Mark in bold, blue color on projects and more with this marker. This indelible, oil-based ink works on all surfaces. Once the ink is dried it presents a semi-permanent finished on Hydrophobic Glass Vials and Inserts. Laboratory Markers Features: Tip Type: Fine Point Marking Width: 1.5mm Marking Temperature Range: -30 ̊ to 60 ̊C Dry Time: 10 Seconds Directions for use: Shake vigorously with cap on. Press the tip on a surface that can be marked until inked. Replace the cap after use.
This item is not returnable. View return policy
Catalog No. 50-227-0499 Supplier Microsolv Technology Corporation Supplier No. 95296MARKOBP
Mark in bold, blue color on projects and more with this marker. This indelible, oil-based ink works on all surfaces. Once the ink is dried it presents a semi-permanent finished on Hydrophobic Glass Vials and Inserts. Laboratory Markers Features: Tip Type: Fine Point Marking Width: 1.5mm Marking Temperature Range: -30 ̊ to 60 ̊C Dry Time: 10 Seconds Directions for use: Shake vigorously with cap on. Press the tip on a surface that can be marked until inked. Replace the cap after use.