Learn More
Med Vet International X-Ray Marker Holder, J1185A
Catalog No. 5000495961
Encompass_Preferred
This device has a malleable arm that holds the x-ray marker at the same height as the bone under investigation. This helps to eliminate the magnification effect of standard radiography.
Catalog No. 50-004-95961 Supplier Med Vet International Supplier No. JOR1185A
This device has a malleable arm that holds the x-ray marker at the same height as the bone under investigation. This helps to eliminate the magnification effect of standard radiography.