Learn More
Grainger MARKAL Paint Marker, Fiber Nib
Catalog No. NC0850762
Encompass_Preferred
The Markal Paint Marker with fiber nib is a paint marker that can write on a variety of surfaces including metal, plastic, glass, rubber, wood, and more. It features oil-based paint ink, has a fiber tip for controlled lines, and is fast drying. This paint marker creates opaque lines and provides good coverage. It is versatile for labeling, marking, writing, and more. The paint marker comes in multiple tip sizes and colors.
Catalog No. NC0850762 Supplier Grainger Supplier No. 2F934
The Markal Paint Marker with fiber nib is a paint marker that can write on a variety of surfaces including metal, plastic, glass, rubber, wood, and more. It features oil-based paint ink, has a fiber tip for controlled lines, and is fast drying. This paint marker creates opaque lines and provides good coverage. It is versatile for labeling, marking, writing, and more. The paint marker comes in multiple tip sizes and colors.