Learn More
Electron Microscopy Sciences Marker Pen Red
SDP
Catalog No. 50949062
Encompass_Preferred
Smear resistant, water-resistant, fast-drying no. 15 fine line. Usable on glass, porcelain, metals, and plastics.
Catalog No. 50-949-062 Supplier Electron Microscopy Sciences Supplier No. 72130
Smear resistant, water-resistant, fast-drying no. 15 fine line. Usable on glass, porcelain, metals, and plastics.