Learn More
Enterprise Technology Solutions Industrial Permanent Marker, Fine Bullet Tip, Black, Dozen
SDP
Catalog No. 502052612
Encompass_Preferred
Sharpie Professional Industrial Permanent Markers feature durable permanent inks that resist fading when put to commercial use. The ink is suited for industrial applications, including extreme heat (up to 500 degrees Fahrenheit). These permanent markers can be used on most materials including metal, glass, plastic, film, and foil.
Catalog No. 50-205-2612 Supplier Enterprise Technology Solutions Supplier No. SAN13601A
Sharpie Professional Industrial Permanent Markers feature durable permanent inks that resist fading when put to commercial use. The ink is suited for industrial applications, including extreme heat (up to 500 degrees Fahrenheit). These permanent markers can be used on most materials including metal, glass, plastic, film, and foil.