Learn More
Essendant Jumbo Refillable Permanent Marker, Broad Chisel Tip, Black
Supplier:  Essendant PIL45100
Encompass_Preferred
When you're faced with big, tough marking and labeling jobs, choose a wide-barrel marker specifically designed for industrial applications. Professional-grade ink delivery system ensures smooth permanent marking on nearly any flat surface: wood, glass, metal, plastic, paper, etc. Specially formulated, low-odor, xylene-free ink dries within seconds. Compatible with Pilot Super Color Marker refill ink (sold separately).
Catalog No. 26-791-07