Learn More
Description
- Constructed of durable clear acrylic
- Universal dispenser works with other pre-packaged items
Specifications
Specifications
| Product Type | Removable General Dispensing Drawer |
| Color | Clear |
| For Use With (Equipment) | Pre-packaged items |
| Material | Acrylic |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.