Learn More
Polysciences, Inc. Marking Dye for Tissue - Black
SDP
Catalog No. NC1929766
Encompass_Preferred
Marking Dyes designed to perfect orientation of excised surgical specimens. This method of permanently marking the margins of tissue specimens is very beneficial to surgeons testing for skin cancer. Can be used for Mohs procedures. Fast drying- no post fixative required. Dyes can be used on fresh or formalin fixed specimens.
Catalog No. NC1929766 Supplier Polysciences, Inc. Supplier No. 241138
Marking Dyes designed to perfect orientation of excised surgical specimens. This method of permanently marking the margins of tissue specimens is very beneficial to surgeons testing for skin cancer. Can be used for Mohs procedures. Fast drying- no post fixative required. Dyes can be used on fresh or formalin fixed specimens.