Learn More
Grainger Marking Flag Blk/Red Elec Plastic PK100
Catalog No. 502752996
Encompass_Preferred
The Grainger Buried Electric Line Marking Flags are used to mark underground electric lines. They are made of durable polyethylene and measure 12 inches tall. The flags feature the words "Buried Electric Line" printed in bold black letters on a bright yellow background for high visibility. The flags slide onto a 36 inch wire stake for easy installation above buried power lines. They help identify the location of underground electric lines to prevent accidental digging or damage. The flags are weather resistant and designed for long term outdoor use.
Catalog No. 50-275-2996 Supplier Grainger Supplier No. 14H738
The Grainger Buried Electric Line Marking Flags are used to mark underground electric lines. They are made of durable polyethylene and measure 12 inches tall. The flags feature the words "Buried Electric Line" printed in bold black letters on a bright yellow background for high visibility. The flags slide onto a 36 inch wire stake for easy installation above buried power lines. They help identify the location of underground electric lines to prevent accidental digging or damage. The flags are weather resistant and designed for long term outdoor use.