Learn More
Brady™ Marking Tape L-Shaped Corner Marks - Adhesive Vinyl, Solid Color, Yellow
Catalog No. 19103852 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Yellow
Catalog No. Color
19-103-852 Yellow
1 options
Catalog No. 19-103-852 Supplier Brady™ Supplier No. 121415
Only null left
Add to Cart
Add to Cart
Permanent acrylic adhesive sticks to a wide variety of surfaces
  • Indoor/outdoor vinyl film withstands exposure to moisture, chemicals and handling.
  • Up to 3 year average outdoor durability
Specifications
Color Yellow
Displayed Text Blank
Length (English) 68.75 ft.
Type Marking Tape
Packaging Quantity 750/Roll
Width (Metric) 114.3 mm
Material Vinyl
Certifications/Compliance ASTM D 2979,NFPA 70E
Adhesive Type Permanent Acrylic