Learn More
Brady™ Marking Tape Roll: High Performance Laminated Vinyl, Checkered, Black on White
Catalog No. 19103773 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black on Yellow
Catalog No. Color
19-103-773 Black on Yellow
1 options
Catalog No. 19-103-773 Supplier Brady™ Supplier No. 76318
  • $124.00
    $82.65 / Each of 1
    Save $41.35
    33% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
High Performance Laminated Vinyl (B-950) tape is perfect for marking obstructions, low hanging objects, pillars, passageways, aisles and other hazardous areas or objects.
  • High gloss, durable vinyl with pressure-sensitive rubber-based adhesive withstands most common chemicals, solvents, oils and water
  • Encapsulated graphics are sealed beneath a protective layer of abrasion-resistant vinyl
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Designed to withstand tough indoor conditions
Specifications
Color Black on Yellow
Packaging Quantity 18 yd./Roll
Width (English) 3 in.
Material Vinyl
Displayed Text Blank
Length (English) 18 yd.
Thickness 0.006 in.
Width (Metric) 100.33 mm
Type Marking Tape