Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Marking Tape Roll: High Performance Laminated Vinyl, Checkered, Black on Yellow

Catalog No. 19103774 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black on Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-774 Black on Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-774 Supplier Brady™ Supplier No. 76319
  • $133.00
    $84.65 / Each of 1

    Save $48.35

    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High Performance Laminated Vinyl (B-950) tape is perfect for marking obstructions, low hanging objects, pillars, passageways, aisles and other hazardous areas or objects.

  • High gloss, durable vinyl with pressure-sensitive rubber-based adhesive withstands most common chemicals, solvents, oils and water
  • Encapsulated graphics are sealed beneath a protective layer of abrasion-resistant vinyl
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Designed to withstand tough indoor conditions

Specifications

Color Black on Yellow
Width (Metric) 100.33 mm
Displayed Text Blank
Packaging Quantity 18 yd./Roll
Length (English) 18 yd.
Thickness 0.006 in.
Type Marking Tape
Width (English) 3 in.
Material Vinyl
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.