Learn More
World Precision Instrument Marmoset Adaptor Kit for 505211
SDP
Catalog No. 502539138
Encompass
  • The Marmoset Adaptor holds the marmoset's head by means of adjustable infra-orbital clamps and a palate bar that is vertically adjustable.
  • The Marmoset Adaptor holdsby means of adjustable infra-orbital clamps and a palate bar that is vertically adjustable with 45 deg ear bars. Fits 505211, 505293, and 505351 U-frame WPI Rat Stereotaxic Instruments
Catalog No. 50-253-9138 Supplier World Precision Instrument Supplier No. 505285
  • The Marmoset Adaptor holds the marmoset's head by means of adjustable infra-orbital clamps and a palate bar that is vertically adjustable.
  • The Marmoset Adaptor holdsby means of adjustable infra-orbital clamps and a palate bar that is vertically adjustable with 45 deg ear bars. Fits 505211, 505293, and 505351 U-frame WPI Rat Stereotaxic Instruments