Learn More
Stoelting™ Marmoset Adaptor
SDP
Catalog No. 10000181 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Adaptor
Catalog No. Model
10-000-181 Adaptor
1 options
Catalog No. 10-000-181 Supplier Stoelting™ Supplier No. 51678
 Restricted Item
Add to Cart
Add to Cart
Holds the marmoset's head by means of adjustable infra-orbital clamps and a palate bar that is vertically adjustable.
Specifications
For Use With (Application) Neuroscience
Animal Marmoset
Model Adaptor