Learn More
Med Vet International Marq-Easy Thermoplastic Splint Sheets, 4" x 6" Sheet, J0940F
Catalog No. 5000581164
Encompass_Preferred
Solid construction 1/8" thick splint sheets offers superior rigidity and tensile strength (180,000 psi), making it ideal for weight-bearing splints in small and large animals (i.e., medium to large dogs, horses, and livestock). The tough, rubberbased material makes a strong, rigid splint, drapes well over contours and bony areas, and adheres to itself when heated to reinforce or patch splint. Radiolucent. Tan.
Catalog No. 50-005-81164 Supplier Med Vet International Supplier No. JOR940F
Solid construction 1/8" thick splint sheets offers superior rigidity and tensile strength (180,000 psi), making it ideal for weight-bearing splints in small and large animals (i.e., medium to large dogs, horses, and livestock). The tough, rubberbased material makes a strong, rigid splint, drapes well over contours and bony areas, and adheres to itself when heated to reinforce or patch splint. Radiolucent. Tan.