Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ MarrowMAX™ Bone Marrow Medium
GREENER_CHOICE

Catalog No. 12260001
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-260-001 500 mL
12-260-014 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-260-001 Supplier Gibco™ Supplier No. 12260001
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fully-supplemented MarrowMAX™ Bone Marrow Medium for short-term culture of bone marrow and other hematopoietic cells for cytogenetic studies and in vitro diagnostic procedures.

Gibco MarrowMAX Bone Marrow Medium is a fully-supplemented medium developed for the short term culture of bone marrow and other hematopoietic cells for cytogenetic studies and in vitro diagnostic procedures.

MarrowMAX Bone Marrow Medium features:

  • Superior performance
  • Ready-to-use format
  • Quality and performance testing
  • Unique, optimized formulation

Superior performance

MarrowMAX Bone Marrow Medium outperforms other commercially available media containing Giant Cell Tumor (GCT) conditioned media, providing consistent performance with a higher mitotic index and superior chromosomal morphology.

Ready-to-use format

MarrowMAX Bone Marrow Medium is a frozen, 1X medium, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2–8°C for up to 60 days.

Quality and performance testing

Every lot of MarrowMAX Bone Marrow Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Bone marrow cells from the aspirate of a normal adult donor are cultured for 24 hours in MarrowMAX Bone Marrow Medium before measuring the mitotic index and chromosome banding resolution. In addition, each lot must pass a sterility test prior to release.

Unique, optimized formulation

MarrowMAX Bone Marrow Medium is an optimized RPMI 1640 medium, supplemented with Fetal Bovine Serum (FBS), gentamicin, and L-glutamine. It also contains a novel human stromal cell conditioned medium that consists of a unique blend of hematopoietic growth factors. MarrowMAX Bone Marrow Medium does not contain GCT conditioned medium.

cGMP Manufacturing and Quality System

MarrowMAX Bone Marrow Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Bone Marrow, Hematopoietic Cell
Concentration 1 X
Form Liquid
Product Type Bone Marrow Medium
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
With Additives Glutamine, Phenol Red, FBS, Conditioned Medium (from Stromal Cells)
Product Line MarrowMAX
Quantity 500 mL
Shipping Condition Dry Ice
Included Antibiotics Gentamicin
Species Human
Content And Storage Storage conditions: -5°C to -20°C. Protect from light.
Shipping conditions: Frozen
Shelf life: 18 months from date of manufacture. Thawed medium can be stored at 2°C to 8°C for up to 60 days.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.