Learn More
Medchemexpress LLC HY-N6995 5mg Medchemexpress, Marrubiin CAS:465-92-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN69955MG
Encompass_Preferred
Medchemexpress, HY-N6995 5mg Marrubiin CAS:465-92-9 Marrubiin, isolated from Marrubium vulgare, exhibits vasorelaxant and antioedematogenic activity. Marrubiin alleviates diabetic symptoms in animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2978